Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566377 Supplier Invitrogen™ Supplier No. PA566377
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLRKRKASPEP PDSAEGALKL GEEQQRQQWM ANQSEALKLT MSAG Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

May contribute to the control of female reproductive function. May play a role in gene transcription by transactivating GNRH1 promoter and repressing PENK promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IRF2BPL
Gene Accession No. Q9H1B7
Gene Alias C14orf4; EAP1; enhanced at puberty 1; enhanced at puberty protein 1; interferon regulatory factor 2 binding protein like; interferon regulatory factor 2 binding protein-like; interferon regulatory factor 2-binding protein-like; IRF2BPL; KIAA1865; My039; Probable E3 ubiquitin-protein ligase IRF2BPL
Gene Symbols IRF2BPL
Host Species Rabbit
Immunogen Recombinant Human C14orf4. Recombinant protein control fragment (Product #RP-105177).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64207
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.