Learn More
C14orf43 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303157AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
ELMSAN1 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include nucleus; DNA binding.Specifications
C14orf43 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q6PJG2 | |
ELMSAN1 | |
Between 550 and 600 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ELMSAN1 | |
c14_5541, C14orf117, C14orf43, chromosome 14 open reading frame 117, chromosome 14 open reading frame 43, ELM2 and Myb/SANT-like domain containing 1, LSR68, MIDEAS | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91748 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.