Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf68 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518647
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518647 Supplier Invitrogen™ Supplier No. PA518647
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene encodes a member of a large family of mitochondrial transporters. The nuclear-encoded carrier protein is embedded in the inner mitochondrial membrane. This member of the family is thought to be an uncoupling protein that uncouples mitochondrial respiration from ATP synthesis by dissipating the transmembrane proton gradient. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf68
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SLC25A47
Gene Accession No. Q6Q0C1
Gene Alias C14orf68; HCC-down-regulated mitochondrial carrier protein; HDMCP; Hepatocellular carcinoma down-regulated mitochondrial carrier protein; hepatocellular carcinoma-downregulated mitochondrial carrier protein; HMFN1655; SLC25A47; Solute carrier family 25 member 47; solute carrier family 25, member 47
Gene Symbols SLC25A47
Host Species Goat
Immunogen Synthetic peptide sequence (RYGNPDAKPTKAD) corresponding to the internal amino acids of C14orf68 (aa 85-98).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283600
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.