Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C14orf80 Monoclonal Antibody (OTI6A3)
GREENER_CHOICE

Catalog No. PIMA527477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527477 Supplier Invitrogen™ Supplier No. MA527477
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C14orf80 Monoclonal Antibody for Western Blot, IHC (P)

C14orf80 may play a role in counteracting perturbation of actin filaments, such as after treatment with the actin depolymerizing microbial metabolite Chivosazole F.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf80
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6A3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TEDC1
Gene Accession No. Q86SX3
Gene Alias C14orf80; chromosome 14 open reading frame 80; TEDC1; Tubulin epsilon and delta complex protein 1; uncharacterized protein C14orf80
Gene Symbols TEDC1
Host Species Mouse
Immunogen Full length human recombinant protein of C14ORF80 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283643
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.