Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C14orf94 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988978
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8978 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8978 Supplier Bioss Supplier No. BS9311R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a subunit of the centrosome complex termed the human augmin complex. The encoded protein localizes to the spindle microtubules and may play a role in mitotic spindle assembly and maintenance of centrosome integrity during cell division. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 1.

Specifications

Antigen C14orf94
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Haus4
Gene Accession No. Q9H6D7
Gene Alias 9430093H08Rik; AU016300; C14orf94; D14Ertd500e; HAUS augmin like complex subunit 4; HAUS augmin-like complex subunit 4; HAUS augmin-like complex, subunit 4; Haus4; homologous to Augmin subunits; RGD1305288
Gene Symbols Haus4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HAUS4/C14orf94.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54930
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.