Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C15orf2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553857
Description
Immunogen sequence: TSVITSKPMN STSVISTVTT NASAHLTSQT AVDPEVVNMD TTAPSQVVIF TSSLSSRVSS LPNSQIHCSA EQRHPGKTSV YTSPLPFIFH NTTPSFNQLF GKEATPQPKF EAPDGQPQKA SLPSA Highest antigen sequence identity to the following orthologs: Mouse - 27%, Rat - 29%.
May be involved in spermatogenesis.
Specifications
C15orf2 | |
Polyclonal | |
Unconjugated | |
NPAP1 | |
C15orf2; NPAP1; nuclear pore associated protein 1; nuclear pore-associated protein 1; protein C15orf2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23742 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NZP6 | |
NPAP1 | |
Recombinant protein corresponding to Human C15orf2. Recombinant protein control fragment (Product #RP-92323). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction