Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C15orf21 Mouse anti-Human, Clone: OTI5A9, lyophilized, TrueMAB™
SDP

Catalog No. 501670191
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-0191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-0191 Supplier OriGene Supplier No. CF810600
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

C15orf21 (HMGN2P46, High Mobility Group Nucleosomal Binding Domain 2 Pseudogene 46) is a Pseudogene. Diseases associated with HMGN2P46 include Prostate Cancer.

Specifications

Antigen C15orf21
Applications Western Blot
Classification Monoclonal
Clone OTI5A9
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene HMGN2P46
Gene Accession No. Q86SG4
Gene Alias C15orf21, D-PCa-2
Gene Symbols HMGN2P46
Host Species Mouse
Immunogen Full length human recombit protein of human C15ORF21 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283651
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.