Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C15orf23 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546284 Supplier Invitrogen™ Supplier No. PA546284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TDTATRRNVR KGYKPLSKQK SEEELKDKNQ LLEAVNKQLH QKLTETQGEL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 85%; Rabbit: 100%; Rat: 79%; Yeast: 82%.

C15orf23 is an essential component of the mitotic spindle required for faithful chromosome segregation and progression into anaphase. It promotes the metaphase-to-anaphase transition and is required for chromosome alignment, normal timing of sister chromatid segregation, and maintenance of spindle pole architecture. The astrin (SPAG5)-kinastrin (SKAP) complex promotes stable microtubule-kinetochore attachments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C15orf23
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KNSTRN
Gene Accession No. Q9Y448
Gene Alias 1700025D04Rik; C10H15orf23; C15orf23; D2Ertd750e; HSD11; Kinastrin; kinetochore localized astrin (SPAG5) binding protein; kinetochore localized astrin/SPAG5 binding protein; kinetochore-localized astrin/SPAG5 binding; kinetochore-localized astrin/SPAG5 binding protein; kinetochore-localized astrin/SPAG5-binding protein; kinetochore-localized astrin-binding protein; Knstrn; putative TRAF4-associated factor 1; SKAP; small kinetochore associated protein; small kinetochore-associated protein; small kinetochore-associated protein, kinetochore-localized astrin-binding protein, TRAF4 associated factor 1; TRAF4 associated factor 1; TRAF4 associated factor 1 homolog; TRAF4AF1; TRAF4-associated factor 1; Unknown (protein for MGC:134039)
Gene Symbols KNSTRN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C15orf23.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90417
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.