Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C15orf23 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735248 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5248 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5248 Supplier Proteintech Group Inc Supplier No. 261891AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C15orf23 is an essential component of the mitotic spindle required for faithful chromosome segregation and progression into anaphase. It promotes the metaphase-to-anaphase transition and is required for chromosome alignment, normal timing of sister chromatid segregation, and maintece of spindle pole architecture. The astrin (SPAG5)-kinastrin (SKAP) complex promotes stable microtubule-kinetochore attachments.

Specifications

Antigen C15orf23
Applications Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KNSTRN
Gene Accession No. Q9Y448
Gene Alias C15orf23, HSD11, KNSTRN, SKAP, TRAF4AF1
Gene Symbols KNSTRN
Host Species Rabbit
Immunogen C15orf23 Fusion Protein Ag21669
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90417
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.