Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C15orf27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559788
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559788 Supplier Invitrogen™ Supplier No. PA559788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLSEDPCPSQ KALDPAPLAR PSPAGSAQTS PELEHRVSLF NQKNQEGFTV FQIRPVIHFQ PTVPMLEDKF RSLESKEQKL HRV Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.

C15orf27 is a voltage-sensor protein present on the post-synaptic side of glutamatergic mossy fibers and granule cells in the cerebellum. Despite the presence of a voltage-sensor segment, it does not form a functional ion channel and its precise role remains unclear. The protein undergoes both rapid and slow structural rearrangements in response to changes in voltage, and contains a zinc-binding site that can regulate the slow conformational transition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C15orf27
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMEM266
Gene Accession No. Q2M3C6
Gene Alias 9630029F15; AI118078; C15orf27; expressed sequence AI118078; HsHVRP1; hTMEM266; Hv1 related protein 1; HV1-related protein 1; HVRP1; Tmem266; Transmembrane protein 266; transmembrane protein C15orf27; transmembrane protein C15orf27 homolog
Gene Symbols TMEM266
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C15orf27. Recombinant protein control fragment (Product #RP-98127).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123591
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.