Learn More
Invitrogen™ C15orf27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113977
Description
Immunogen sequence: EMEMVIQQYE KAKVIQDEQL ERLTQICQEQ GFEIRQLRAH LAQQDLDLAA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Goat: 90%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
C15orf27 is a voltage-sensor protein present on the post-synaptic side of glutamatergic mossy fibers and granule cells in the cerebellum. Despite the presence of a voltage-sensor segment, it does not form a functional ion channel and its precise role remains unclear. The protein undergoes both rapid and slow structural rearrangements in response to changes in voltage, and contains a zinc-binding site that can regulate the slow conformational transition.
Specifications
| C15orf27 | |
| Polyclonal | |
| Unconjugated | |
| TMEM266 | |
| 9630029F15; AI118078; C15orf27; expressed sequence AI118078; HsHVRP1; hTMEM266; Hv1 related protein 1; HV1-related protein 1; HVRP1; Tmem266; Transmembrane protein 266; transmembrane protein C15orf27; transmembrane protein C15orf27 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 123591 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q2M3C6 | |
| TMEM266 | |
| Synthetic peptide directed towards the middle region of human C15orf27. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.