Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C15orf29 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 247951AP150UL

Description
The function of the C15orf29 protein has not yet been determined.Specifications
C15orf29 | |
Polyclonal | |
Unconjugated | |
KATNBL1 | |
C15orf29, Katanin p80 subunit B-like 1, KATNB1-like protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
691543, 72425, 79768 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CWJ3, Q9H079 | |
Katnbl1 | |
C15orf29 Fusion Protein Ag20700 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction