Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C15orf39 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565318
Description
Immunogen sequence: TPRCPLDFAPQ TLSFPYARDD LSLYGASPGL GGTPPSQNNV RAVPQPGAFQ RACQPLPASQ PCSEPVRPAQ EAEEKTWLPS CRKEKLQPRL SEHSG Highest antigen sequence identity to the following orthologs - mouse 80%, rat 78%.
C15orf39 is a protein coding gene.
Specifications
C15orf39 | |
Polyclonal | |
Unconjugated | |
C15orf39 | |
C15orf39; chromosome 15 open reading frame 39; FP6578; Uncharacterized protein C15orf39 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56905 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZRI6 | |
C15orf39 | |
Recombinant Human C15orf39. Recombinant protein control fragment (Product #RP-104745). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction