Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C15orf40 Antibody (OTI2B7), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272375H
Description
C15orf40 Monoclonal specifically detects C15orf40 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
C15orf40 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
C15orf40 | |
Full length recombinant protein of human C15orf40 (NP_653198) produced in HEK293T cells. | |
0.1 mL | |
123207.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2B7 | |
Western Blot | |
chromosome 15 open reading frame 40, FLJ33606, hypothetical protein LOC123207, MGC29937 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction