Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C15orf40 Monoclonal Antibody (OTI1D8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501600S


Description
C15orf40 Monoclonal antibody specifically detects C15orf40 in Human samples. It is validated for Western BlotSpecifications
C15orf40 | |
Monoclonal | |
0.61 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P34913 | |
Mouse | |
Affinity chromatography | |
RUO | |
2053 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI1D8 | |
Unconjugated | |
EPHX2 | |
3110040N11Rik; AI851475; C15orf40; chromosome 15 open reading frame 40; RIKEN cDNA 3110040N11 gene; UPF0235 protein C15orf40; UPF0235 protein C15orf40 homolog | |
Full length human recombinant protein of human EPHX2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction