Learn More
C15orf40 Monoclonal Antibody (OTI1D9), TrueMAB™, OriGene

Description
Specifications
Specifications
| Antigen | C15orf40 |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI1D9 |
| Concentration | 0.52 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | HID1 |
| Gene Accession No. | Q8IV36 |
| Gene Alias | 3110040N11Rik; AI851475; C15orf40; chromosome 15 open reading frame 40; RIKEN cDNA 3110040N11 gene; UPF0235 protein C15orf40; UPF0235 protein C15orf40 homolog |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.