Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C15orf48 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582476 Supplier Invitrogen™ Supplier No. PA582476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDVILDRKKN PEPWETVDPT VPQKLITINQ QWKPIEELQ Highest antigen sequence indentity to the following orthologs: Rat - 79%, Mouse - 85%.

This gene was first identified in a study of human esophageal squamous cell carcinoma tissues. Levels of both the message and protein are reduced in carcinoma samples. In adult human tissues, this gene is expressed in the esophagus, stomach, small intestine, colon and placenta. Alternatively spliced transcript variants that encode the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C15orf48
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C15orf48
Gene Accession No. Q9C002
Gene Alias C15orf48; chromosome 15 open reading frame 48; FOAP-11; NMES1; normal mucosa of esophagus specific 1; normal mucosa of esophagus-specific gene 1 protein; Protein FOAP-11
Gene Symbols C15orf48
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NMES1. Recombinant protein control fragment (Product #RP-89863).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84419
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.