Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C15orf59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571037
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571037 Supplier Invitrogen™ Supplier No. PA571037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 79%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rat: 100%.

Component of the protein machinery at the inhibitory synapses, probably acting as a scaffold. Inhibitory synapses dampen neuronal activity through postsynaptic hyperpolarization. This synaptic inhibition is fundamental for the functioning of the central nervous system, shaping and orchestrating the flow of information through neuronal networks to generate a precise neural code.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C15orf59
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C15orf59
Gene Accession No. Q2T9L4
Gene Alias C15orf59; chromosome 15 open reading frame 59; Inhibitory synaptic factor 1; INSYN1; UPF0583 protein C15orf59
Gene Symbols C15orf59
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human C15orf59.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388135
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.