Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C16orf57 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537975
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537975 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537975 Supplier Invitrogen™ Supplier No. PA537975
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is expected to recognize both reported isoforms (NP_078874.2; NP_001182231.1). It is predicted to react with pig based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

C16orf57 is a phosphodiesterase responsible for the U6 snRNA 3' end processing. This protein acts as an exoribonuclease (RNase) responsible for trimming the poly(U) tract of the last nucleotides in the pre-U6 snRNA molecule, leading to the formation of mature U6 snRNA 3' end-terminated with a 2',3'-cyclic phosphate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C16orf57
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene USB1
Gene Accession No. Q5I0I5, Q91W78, Q9BQ65
Gene Alias AA960436; C16orf57; FLJ13154; HAMAP-Rule:MF_03040}; hUsb1; HVSL motif containing 1; HVSL1; Mpn1; mutated in poikiloderma with neutropenia protein 1; PN; putative U6 snRNA phosphodiesterase; RGD1305215; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA biogenesis phosphodiesterase 1; U6 snRNA phosphodiesterase; U6 snRNA phosphodiesterase {ECO:0000255; UPF0406 protein C16orf57; UPF0406 protein C16orf57 homolog; USB1
Gene Symbols USB1
Host Species Goat
Immunogen Peptide with sequence C-TANQVKIYTNQEKT, from the internal region of C16orf57 (aa154-167).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101985, 307643, 79650
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.