Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C16orf57 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9631R

Description
C16orf57 is a phosphodiesterase responsible for the U6 snRNA 3′ end processing. This protein acts as an exoribonuclease (RNase) responsible for trimming the poly(U) tract of the last nucleotides in the pre-U6 snRNA molecule, leading to the formation of mature U6 snRNA 3′ end-terminated with a 2′,3′-cyclic phosphate.Specifications
C16orf57 | |
Polyclonal | |
Unconjugated | |
USB1 | |
AA960436; C16orf57; FLJ13154; HAMAP-Rule:MF_03040}; hUsb1; HVSL motif containing 1; HVSL1; Mpn1; mutated in poikiloderma with neutropenia protein 1; PN; putative U6 snRNA phosphodiesterase; RGD1305215; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA biogenesis phosphodiesterase 1; U6 snRNA phosphodiesterase; U6 snRNA phosphodiesterase {ECO:0000255; UPF0406 protein C16orf57; UPF0406 protein C16orf57 homolog; USB1 | |
Rabbit | |
Protein A | |
RUO | |
79650 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9BQ65 | |
USB1 | |
KLH conjugated synthetic peptide derived from human C16orf57. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction