Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C16orf58 Antibody, Novus Biologicals™
SDP

Catalog No. p-7108210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159541 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159541 Supplier Novus Biologicals Supplier No. NBP159541
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C16orf58 Polyclonal specifically detects C16orf58 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen C16orf58
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96GQ5
Gene Alias chromosome 16 open reading frame 58, FLJ13868, hypothetical protein LOC64755
Gene Symbols C16ORF58
Host Species Rabbit
Immunogen Synthetic peptides corresponding to C16ORF58 The peptide sequence was selected from the N terminal of C16ORF58. Peptide sequence QAVFLPQGFPDSVSPDYLPYQLWDSVQAFASSLSGSLATQAVLLGIGVGN.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64755
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Human: 100%; Pig: 100%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.