Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C16orf59 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584253 Supplier Invitrogen™ Supplier No. PA584253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EKAVRVRRGI TKAGERDKAP SLKSRSIVTS SGTTASAPPH SPGQAGGHAS DTRPTKGLRQ TTVPAKGHPE RRLLSVGDGT RVGM Highest antigen sequence indentity to the following orthologs: Mouse - 45%, Rat - 54%.

C16orf59 (TEDC2, Tubulin Epsilon And Delta Complex 2) is a Protein Coding gene. Acts as a positive regulator of ciliary hedgehog signaling. Required for centriole stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C16orf59
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TEDC2
Gene Accession No. Q7L2K0
Gene Alias C16orf59; chromosome 16 open reading frame 59; FLJ13909; TEDC2; Tubulin epsilon and delta complex protein 2; uncharacterized protein C16orf59
Gene Symbols TEDC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C16orf59. Recombinant protein control fragment (Product #RP-108410).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80178
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.