Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C16orf65 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570140 Supplier Invitrogen™ Supplier No. PA570140
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 77%; Human: 100%; Rabbit: 92%; Rat: 77%.

PDZD9 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C16orf65
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PDZD9
Gene Accession No. Q8IXQ8
Gene Alias 4930408O21Rik; C16orf65; LRRGT00105; PDZ domain containing 9; PDZ domain-containing protein 9; PDZ domain-containing protein C16orf65; PDZ domain-containing protein C16orf65 homolog; PDZD9
Gene Symbols PDZD9
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C16orf65.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255762
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.