Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C16orf72 Antibody (OTI2D1), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271842APC
Description
C16orf72 Monoclonal specifically detects C16orf72 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
| C16orf72 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| C16orf72 | |
| Full length recombinant protein of human C16orf72 (NP_054836) produced in HEK293T cells. | |
| 0.1 mL | |
| 29035.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2D1 | |
| Flow Cytometry | |
| chromosome 16 open reading frame 72, FLJ41272, hypothetical protein LOC29035, PRO0149 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction