Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C16orf72 Antibody (OTI2D1), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271842JF646
Description
C16orf72 Monoclonal specifically detects C16orf72 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
C16orf72 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C16orf72 | |
Full length recombinant protein of human C16orf72 (NP_054836) produced in HEK293T cells. | |
0.1 mL | |
29035.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI2D1 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
chromosome 16 open reading frame 72, FLJ41272, hypothetical protein LOC29035, PRO0149 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction