Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C16orf72 Antibody (OTI2D1), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271842MFV610
Description
C16orf72 Monoclonal antibody specifically detects C16orf72 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
C16orf72 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
29035 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI2D1 | |
Flow Cytometry | |
chromosome 16 open reading frame 72, FLJ41272, hypothetical protein LOC29035, PRO0149 | |
Full length human recombinant protein of human C16orf72 (NP_054836) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction