Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C16orf72 Monoclonal Antibody (OTI1A2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501562S


Description
C16orf72 Monoclonal antibody specifically detects C16orf72 in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
C16orf72 | |
Monoclonal | |
0.79 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P49757 | |
Mouse | |
Affinity chromatography | |
RUO | |
8650 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunocytochemistry, Western Blot | |
OTI1A2 | |
Unconjugated | |
Numb | |
C16orf72; chromosome 16 open reading frame 72; PRO0149; UPF0472 protein C16orf72 | |
Full length human recombinant protein of human NUMB produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction