Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C16orf72 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559626
Description
Immunogen sequence: KLWHLFQNSA TAVAQLYKDR VCQQPGLSLW VPFQNAATAV TNLYKESVDT HQRSFDIGIQ IGYQRRNKDV LAWVKKRRRT IRREDLISFL C Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
C16orf72 is a protein coding gene.
Specifications
C16orf72 | |
Polyclonal | |
Unconjugated | |
C16orf72 | |
C16orf72; chromosome 16 open reading frame 72; PRO0149; UPF0472 protein C16orf72 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14CZ0 | |
C16orf72 | |
Recombinant protein corresponding to Human C16orf72. Recombinant protein control fragment (Product #RP-98783). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction