Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C16orf72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566928 Supplier Invitrogen™ Supplier No. PA566928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRSSTPGSPTH VSSGSNASRR RNGLHDVDLN TFISEEMALH LDNGGTRKRT SAQCGDVITD SPT Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

C16orf72 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C16orf72
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C16orf72
Gene Accession No. Q14CZ0
Gene Alias C16orf72; chromosome 16 open reading frame 72; PRO0149; UPF0472 protein C16orf72
Gene Symbols C16orf72
Host Species Rabbit
Immunogen Recombinant Human C16orf72. Recombinant protein control fragment (Product #RP-107580).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.