Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C16orf87 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598546
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598546 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598546 Supplier Invitrogen™ Supplier No. PA598546
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C16orf87 Polyclonal Antibody for Western Blot, IP, ELISA

C16orf87 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C16orf87
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene C16orf87
Gene Accession No. Q6PH81
Gene Alias C16orf87; chromosome 16 open reading frame 87; UPF0547 protein C16orf87
Gene Symbols C16orf87
Host Species Rabbit
Immunogen Recombinant Human UPF0547 protein C16orf87 protein (1-154AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388272
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.