Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C16orf89 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553071
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553071 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553071 Supplier Invitrogen™ Supplier No. PA553071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSVREKWAQE PLLQPLSLRV GMLGEKLEAA IQRSLHYLKL SDPKYLREFQ LTLQPGFWKL PHAWIHTDAS LVYPTFGPQD SFSEERSDVC LVQLLGTGTD SSEPCGLSDL CRSLMTKPGC SGYCLSHQLL FFLWA Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 73%.

This gene is expressed predominantly in the thyroid. Based on expression patterns similar to thyroid transcription factors and proteins, this gene may function in the development and function of the thyroid. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Oct 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C16orf89
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C16orf89
Gene Accession No. Q6UX73
Gene Alias C16orf89; chromosome 16 open reading frame 89; MGC45438; UNQ904/PRO1925; UPF0764 protein C16orf89
Gene Symbols C16orf89
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C16orf89. Recombinant protein control fragment (Product #RP-89929).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146556
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.