Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C16orf93 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559740 Supplier Invitrogen™ Supplier No. PA559740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SCLPQFAFFP QPPLPRPRIC MWKYLDVHSM HQLEKTTNAE MREVLAELLE LGCPEQSLRD AITLDLFCHA LIFCRQQGFS L Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.

C16orf93, also named as CCDC189, is a coiled-coil domain-containing protein which has four isoforms with MW 38 kDa, 34 kDa, 12 kDa and 20 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C16orf93
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC189
Gene Accession No. A1A4V9
Gene Alias C16orf93; CCDC189; coiled-coil domain containing 189; coiled-coil domain-containing protein 189
Gene Symbols CCDC189
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C16orf93. Recombinant protein control fragment (Product #RP-98590).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90835
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.