Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C16orf95 Polyclonal Antibody
C16orf95 Polyclonal Antibody
Supplier: Thermo Scientific PA559324
Description
C16orf95 Polyclonal Antibody for IHC

Specifications
C16orf95 | |
Polyclonal | |
Unconjugated | |
C16orf95 | |
C16orf95; chromosome 16 open reading frame 95; uncharacterized protein C16orf95 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100506581 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H693 | |
C16orf95 | |
Recombinant protein corresponding to Human C16orf95. Recombinant protein control fragment (Product #RP-98637). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction