Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf104 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570024 Supplier Invitrogen™ Supplier No. PA570024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

Is required for meiosis completion in both male and female germ cells. Confers stability to numerous meiotic mRNAs in gonads allowing proper initiation and progression into meiosis prophase I. The function may involve YTHDC2 and is independent of induction by retinoic acid (RA). Maintains an extended meiotic prophase I by properly promoting the transition from a mitotic to a meiotic cell cycle program by binding transcripts through its interaction with YTHDC2 that regulate the mitotic cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf104
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MEIOC
Gene Accession No. A2RUB1
Gene Alias C11H17orf104; C17orf104; C19H17orf104; C9H17orf104; Gm1564; Gm663; MEIOC; meiosis specific with coiled-coil domain; meiosis-specific coiled-coil domain-containing protein MEIOC; Meiosis-specific with coiled-coil domain protein; predicted gene 1564; RGD1563746; similar to nuclear protein with a coiled coil-4 domain of bilaterial origin like (3L720); uncharacterized protein C17orf104; uncharacterized protein C17orf104 homolog
Gene Symbols MEIOC
Host Species Rabbit
Immunogen synthetic peptide directed towards the C terminal region of human C17orf104.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284071
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.