Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CFAP97D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571012 Supplier Invitrogen™ Supplier No. PA571012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 86%.

CFAP97D1 is a protein that is encoded by the CFAP97D1 gene in humans. It is a component of the cilia and flagella, which are hair-like structures that protrude from the surface of cells and are involved in various cellular processes, including movement and signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CFAP97D1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C17orf105
Gene Accession No. B2RV13
Gene Alias C17orf105; CFAP97 domain-containing protein 1; CFAP97D1; chromosome 17 open reading frame 105; uncharacterized protein C17orf105; Uncharacterized protein CFAP97D1
Gene Symbols C17orf105
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C17orf105.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284067
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.