Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554472 Supplier Invitrogen™ Supplier No. PA554472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ENQKILTPLV SLDTPGKATV QVVILADPDG HEICFVGDEA FRELSKMDPE GSKLLDDAMA ADKSDEWFAK HNKPKASG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.

GLOD4 (Glyoxalase Domain Containing 4) is a Protein Coding gene. An important paralog of this gene is GLO1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf25
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLOD4
Gene Accession No. Q5I0D1, Q9CPV4, Q9HC38
Gene Alias 1700082G03Rik; 2700085E05Rik; C17orf25; C81254; CGI-150; Glod4; glyoxalase domain containing 4; glyoxalase domain-containing protein 4; HC71; My027; RGD1307010
Gene Symbols GLOD4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C17orf25. Recombinant protein control fragment (Product #RP-93380).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363644, 51031, 67201
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.