Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf28 Monoclonal Antibody (OTI2G6), TrueMAB™, OriGene
SDP

Catalog No. 502542340
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2340 30 μL
50-167-2514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2340 Supplier OriGene Supplier No. TA501371S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C17orf28 Monoclonal antibody specifically detects C17orf28 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen C17orf28
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2G6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HID1
Gene Accession No. Q8IV36
Gene Alias C17orf28; C630004H02Rik; C9H17orf28; DMC1; downregulated in multiple cancer 1; Down-regulated in multiple cancers 1; Hid1; HID-1; HID1 domain containing; HID1 domain-containing protein; protein HID1; Protein hid-1 homolog; RGD1311422; UPF0663 transmembrane protein C17orf28; UPF0663 transmembrane protein C17orf28 homolog
Host Species Mouse
Immunogen Full length human recombinant protein of human C17orf28 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283987
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.