Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf28 Mouse anti-Human, Clone: OTI2G6, lyophilized, TrueMAB™
SDP

Catalog No. 501660390
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-0390 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-0390 Supplier OriGene Supplier No. CF501371
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

CQ028 may play an important role in the development of cancers in a broad range of tissues.

Specifications

Antigen C17orf28
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone OTI2G6
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene HID1
Gene Accession No. Q8IV36
Gene Alias 17orf28, C17orf28, DMC1, HID-1
Gene Symbols HID1
Host Species Mouse
Immunogen Full length human recombit protein of human C17orf28 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283987
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.