Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf37 Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene
SDP

Catalog No. 502540214
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0214 30 μL
50-167-5266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0214 Supplier OriGene Supplier No. TA504614S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C17orf37 Monoclonal antibody specifically detects C17orf37 in Human samples. It is validated for Western Blot

Specifications

Antigen C17orf37
Applications Western Blot
Classification Monoclonal
Clone OTI1G2
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MIEN1
Gene Accession No. Q9BRT3
Gene Alias 1810046J19Rik; AI463380; C16H17orf37; C17orf37; C35; HBV XAg-transactivated protein 4; HBV X-transactivated gene 4 protein; MGC14832; MIEN1; Migration and invasion enhancer 1; ORB3; protein C17orf37; protein C17orf37 homolog; Protein C35; RDX12; RGD1306682; XTP4
Host Species Mouse
Immunogen Full length human recombinant protein of human C17orf37 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84299
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.