Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADPRM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554740
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554740 Supplier Invitrogen™ Supplier No. PA554740
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSSPKYEQCM KILREHNPNT ELNSPQGLSE PQFVQFNGGF SQEQLNWLNE VLTFSDTNQE KVVIVSHLPI YP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

ADPRM (ADP-ribose/CDP-alcohol diphosphatase) is a protein that belongs to the Nudix hydrolase family. It plays a crucial role in the metabolism of nucleotide sugars and nucleotide sugar alcohols, which are important for glycosylation reactions in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADPRM
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADPRM
Gene Accession No. Q3LIE5
Gene Alias 2310004I24Rik; ADPRibase-Mn; ADP-ribose/CDP-alcohol diphosphatase, manganese dependent; ADP-ribose/CDP-alcohol diphosphatase, manganese-dependent; ADP-ribose/CDP-alcohol pyrophosphatase; Adprm; C17orf48; CDP-choline phosphohydrolase; liver manganese (II)-dependent ADP-ribose/CDP-alcohol pyrophosphatase; manganese-dependent ADP-ribose/CDP-alcohol diphosphatase; Manganese-dependent ADP-ribose/CDP-alcohol diphosphatase-like protein; MDS006; NBLA03831; RGD1309906
Gene Symbols ADPRM
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C17orf48. Recombinant protein control fragment (Product #RP-93419).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.