Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADPRM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554893
Description
Immunogen sequence: ALAVIWSHEC VVCFFAGHTH DGGYSEDPFG VYHVNLEGVI ETAPDSQAFG TVHVYPDKMM LKGRGRVPDR IMNYKKERAF HC Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 85%.
ADPRM (ADP-ribose/CDP-alcohol diphosphatase) is a protein that belongs to the Nudix hydrolase family. It plays a crucial role in the metabolism of nucleotide sugars and nucleotide sugar alcohols, which are important for glycosylation reactions in cells.
Specifications
ADPRM | |
Polyclonal | |
Unconjugated | |
ADPRM | |
2310004I24Rik; ADPRibase-Mn; ADP-ribose/CDP-alcohol diphosphatase, manganese dependent; ADP-ribose/CDP-alcohol diphosphatase, manganese-dependent; ADP-ribose/CDP-alcohol pyrophosphatase; Adprm; C17orf48; CDP-choline phosphohydrolase; liver manganese (II)-dependent ADP-ribose/CDP-alcohol pyrophosphatase; manganese-dependent ADP-ribose/CDP-alcohol diphosphatase; Manganese-dependent ADP-ribose/CDP-alcohol diphosphatase-like protein; MDS006; NBLA03831; RGD1309906 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3LIE5 | |
ADPRM | |
Recombinant protein corresponding to Human C17orf48. Recombinant protein control fragment (Product #RP-93420). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction