Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf49 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555083
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555083 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555083 Supplier Invitrogen™ Supplier No. PA555083
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESPKKGPKKV ASGVLSPPPA APPPSSSSVP EAGGPPIKKQ KADVTLSALN DSDANSDVVD IEGLGETPPA KKLNFD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

Component of chromatin complexes such as the MLL1/MLL and NURF complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf49
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C17orf49
Gene Accession No. Q8IXM2
Gene Alias 0610010K14Rik; 1110020A23Rik; AL033328; AU045833; BAP18; BPTF associated protein of 18 kDa; BPTF-associated protein of 18 kDa; C17orf49; Chromatin complexes subunit BAP18; chromosome 17 open reading frame 49; HEPIS; human embryo lung cellular protein interacting with SARS-CoV nsp-10; MLL1/MLL complex subunit C17orf49; MLL1/MLL complex subunit C17orf49 homolog; RIKEN cDNA 0610010K14 gene
Gene Symbols C17orf49
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C17orf49. Recombinant protein control fragment (Product #RP-93367).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124944
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.