Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf49 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989222
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9222 Supplier Bioss Supplier No. BS9640R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Component of chromatin complexes such as the MLL1/MLL and NURF complexes.

Specifications

Antigen C17orf49
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C17orf49
Gene Accession No. Q8IXM2
Gene Alias BAP18; BPTF associated protein of 18 kDa; BPTF-associated protein of 18 kDa; C17orf49; Chromatin complexes subunit BAP18; chromosome 17 open reading frame 49; HEPIS; human embryo lung cellular protein interacting with SARS-CoV nsp-10; MLL1/MLL complex subunit C17orf49
Gene Symbols C17orf49
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C17orf49.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124944
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.