Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C17orf58 Antibody (OTI6E9), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272489AF700
Description
C17orf58 Monoclonal specifically detects C17orf58 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
C17orf58 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C17orf58 | |
Full length recombinant protein of human C17orf58 (NP_858041) produced in E.coli. | |
0.1 mL | |
284018.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI6E9 | |
Western Blot | |
chromosome 17 open reading frame 58, hypothetical protein LOC284018, MGC138278 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction