Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf62 Monoclonal Antibody (OTI3F10)
GREENER_CHOICE

Catalog No. PIMA526302
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526302 Supplier Invitrogen™ Supplier No. MA526302
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C17orf62 Monoclonal Antibody for Western Blot, ICC/IF

C17orf62 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf62
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3F10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene C17ORF62
Gene Accession No. Q9BQA9
Gene Alias C17orf62; chromosome 17 open reading frame 62; CYBC1; Cytochrome b-245 chaperone 1; EROS; Essential for reactive oxygen species protein; HAPSTR1; uncharacterized protein C17orf62
Gene Symbols C17ORF62
Host Species Mouse
Immunogen Full length human recombinant protein of C17orf62 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79415
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.