Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C17orf64 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113071
Description
C17orf64 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| C17orf64 | |
| Polyclonal | |
| Unconjugated | |
| C17orf64 | |
| C17orf64; chromosome 17 open reading frame 64; uncharacterized protein C17orf64 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 124773 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q86WR6 | |
| C17orf64 | |
| Recombinant Human Uncharacterized protein C17orf64 protein (111-236AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction