Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C17orf66 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9226 Supplier Bioss Supplier No. BS9644R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HEATR9 gene ontology annotations related to this gene include hematopoietic progenitor cell differentiation.

Specifications

Antigen C17orf66
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HEATR9
Gene Accession No. A2RTY3
Gene Alias C17orf66; C9H17orf66; HEAT repeat containing 9; HEAT repeat-containing protein 9; HEATR9; Protein HEATR9; RGD1559875
Gene Symbols HEATR9
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C17orf66.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 256957
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.