Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C17orf70 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555334
Description
Immunogen sequence: VLCSVSPSGS RVPHDLLGGS GGFTLEDALF GLLFGADATL LQSPVVLCGL PDGQLCCVIL KALVTSRSAP GDPNALVKIL HHLEEPVIFI GALKTEPQAA EAAENFLPDE DVHCDC Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.
FAAP100 is a component of the Fanconi anemia (FA; MIM 277650) core complex and is required for core complex stability and FANCD2 (see MIM 227646) monoubiquitination (Ling et al., 2007 [PubMed 17396147]).
Specifications
C17orf70 | |
Polyclonal | |
Unconjugated | |
FAAP100 | |
2310003H01Rik; C17orf70; FAAP100; Fanconi anemia associated protein 100 kDa subunit; Fanconi anemia core complex 100 kDa subunit; Fanconi anemia core complex associated protein 100; Fanconi anemia core complex-associated protein 100; Fanconi anemia-associated protein of 100 kDa; Fanconi anemia-associated protein, 100kDa; RGD1306926; RIKEN cDNA 2310003H01 gene; similar to hypothetical protein FLJ22175 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80233 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0VG06 | |
FAAP100 | |
Recombinant protein corresponding to Human C17orf70. Recombinant protein control fragment (Product #RP-93510). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction