Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf70 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530832
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530832 Supplier Invitrogen™ Supplier No. PA530832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, K562, HL-60, NCI-H929. Predicted reactivity: Human (99%), Rhesus Monkey (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

FAAP100 is a component of the Fanconi anemia (FA; MIM 277650) core complex and is required for core complex stability and FANCD2 (see MIM 227646) monoubiquitination (Ling et al., 2007 [PubMed 17396147]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf70
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene FAAP100
Gene Accession No. Q0VG06
Gene Alias 2310003H01Rik; C17orf70; FAAP100; Fanconi anemia associated protein 100 kDa subunit; Fanconi anemia core complex 100 kDa subunit; Fanconi anemia core complex associated protein 100; Fanconi anemia core complex-associated protein 100; Fanconi anemia-associated protein of 100 kDa; Fanconi anemia-associated protein, 100kDa; RGD1306926; RIKEN cDNA 2310003H01 gene; similar to hypothetical protein FLJ22175
Gene Symbols FAAP100
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FAAP100. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80233
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.