Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C17orf71 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555202 Supplier Invitrogen™ Supplier No. PA555202
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SINCLFTVPA NQAFVYIVPG SQEEDPVGML LDQLRSHCTV KDPESLLVPA PLSGPRRYQV MRQHSRQQLS FHIDSSSSSS SGQLVDFTLR EFLWQHVELV LSKKGFDDSV GRNPQPSHFE LPTYQKWISA ASK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Involved in nonsense-mediated decay (NMD) of mRNAs containing premature stop codons. Is recruited by release factors to stalled ribosomes together with SMG1 and SMG9 (forming the SMG1C protein kinase complex) and, in the SMG1C complex, is required to mediate the recruitment of SMG1 to the ribosome:SURF complex and to suppress SMG1 kinase activity until the ribosome:SURF complex locates the exon junction complex (EJC). Acts as a regulator of kinase activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf71
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SMG8
Gene Accession No. Q8ND04
Gene Alias 1200011M11Rik; ABC2; Amplified in breast cancer gene 2 protein; C17orf71; C19H17orf71; CG6729 gene product; FLJ10587; FLJ23205; Protein SMG8; protein smg-8 homolog; RGD1306862; Smg8; smg-8 homolog, nonsense mediated mRNA decay factor; smg-8 homolog, nonsense mediated mRNA decay factor (C. elegans); SMG8 nonsense mediated mRNA decay factor; SMG8, nonsense mediated mRNA decay factor
Gene Symbols SMG8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C17orf71. Recombinant protein control fragment (Product #RP-93372).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.